Cambia visualizzazione
| Voi/SKU | Disponibilità | Prezzo |
|---|
Informazioni su questo articolo
NACRES:
NB.51
UNSPSC Code:
41121800
eCl@ss:
32039090
Prezzi e disponibilità al momento non sono disponibili
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutartipackaging
pkg of 1 ea
manufacturer/tradename
Synthware C1301
suitability
suitable for chromatography
General description
0-4 mm high vacuum valve with metering tip to adjust air pressure. With standard taper joint. Order CC3525 pinch type clamps with screw-locking device for ball joints. For replacement O-rings on high vacuum plug shaft, order R010. The O-rings on the high vacuum PTFE valve are completely encapsulated with thin layer of PTFE, making it inert to chemicals.
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
Questo articolo | |||
|---|---|---|---|
| description with PTFE stopcock | description joint: ST/NS 24/40, with PTFE stopcock | description hose connection O.D. 16 mm | description hose connection O.D. 10 mm |
| manufacturer/tradename Synthware C1301 | manufacturer/tradename Synthware C1324 | manufacturer/tradename Synthware M131605 | manufacturer/tradename Synthware M131604 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| suitability suitable for chromatography | suitability suitable for chromatography | suitability - | suitability - |
Still not finding the right product?
Explore all of our products under Synthware™ chromatography flow control adapter
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?