Vai a
BCS-262CS
CoolCell™ SV10
Stem Cell Cryopreservation System
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Informazioni su questo articolo
UNSPSC Code:
41103000
feature
for vials 10 mL, Includes XT Starter cooling base with CoolRack SV10 and CoolCell SV10 cell freezing container
manufacturer/tradename
BioCision BCS-262CS
General description
CoolCell SV10 Stem Cell Cryopreservation System is a kit designed for efficient and controlled preparation and cryopreservation of stem cells and cell therapy products.
Ice-free cell preparation:
The XT Starter ice-free cooling base and CoolRack SV10 vial module provide up to 4 hours of ice-free cooling (0.5 to 4.0°C) for cell and cryoprotectant preparation. Simply freeze the XT Starter core in a -20°C freezer then rest the thermo-conductive CoolRack SV10 on top. The system will equilibrate to approximately 2°C and keep all samples uniformly cold and free of ice contamination. The small footprint and low profile make this an ideal cooling solution for cell preparation performed in a tissue culture a hood.
Dimensions:
XT Starter ice-free cooling base: (L x W x H): 16.2 x 12.0 x 5.5 cm /CoolRack SV10 module: (L x W x H): 12.7 x 8.6 x 3.8 cm; Well dimensions: (Diam x height): 23.6 x 48 mm; Holds 12 vials.
Controlled-rate cell freezing:
The CoolCell SV10 cell freezing container provides highly reproducible -1°C/minute freezing rate for cells when cryopreserved in a -80°C freezer. No programming or alcohol is required - simply place in a -80°C freezer. The insulative outer housing and thermo-conductive core ensure consistent heat removal to all vials throughout the freezing period. CoolCell containers are unbreakable, open easily when frozen, and are not cold to the touch when frozen - no "frosty" fingers.
Holds 6 injectable vials.
Legal Information
CoolCell is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)