Vai a
MTCD309645
MTC™ Bio OptiWell™ Deep Well Plate
96 square wells, V-bottom wells, capacity 2.3 mL, non-sterile, case of 50 ea (10 bags of 5ea)
Sinonimo/i:
automated research plate, MTC Bio D3096-45, automated assay plate, lab automation plate, research automation tray, robotic research plate
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
Informazioni su questo articolo
UNSPSC Code:
41121800
Material:
V-bottom wells, clear plate, clear wells, polypropylene , square wells
Size:
96 wells
Sterility:
non-sterile
Feature:
lid: no, skirt: no
Prezzi e disponibilità al momento non sono disponibili
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
V-bottom wells, clear plate, clear wells, polypropylene , square wells
sterility
non-sterile
feature
lid: no, skirt: no
packaging
case of 50 ea (10 bags of 5ea)
manufacturer/tradename
MTC™ Bio D3096-45
parameter
4,000 × g speed
capacity
2.3 mL
size
96 wells
well volume
2.3 mL
General description
OptiWell plates come in 24, 48, 96 or 384 format for virtually every research lab, HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation and all are certified non-pyrogenic, RNase/DNase free.
Features and Benefits
  • For automated research platforms / robotic equipment
  • Produced/packaged in a Class 100,000 clean room
  • ANSI / SLAS 1-2004 compliant
  • Minimal binding / liquid retention
  • Black printed alpha-numerics
  • Virgin, medical-grade polypropylene
  • Sterilized by gamma irradiation
  • Certified non-pyrogenic, RNase/DNase free
Legal Information
MTC is a trademark of MTC Bio
OptiWell is a trademark of MTC Bio
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
MTCD309645SMTCD309643MTCD309637
description
96 square wells, V-bottom wells, capacity 2.3 mL, non-sterile, case of 50 ea (10 bags of 5ea)
description
96 square wells, V-bottom wells, capacity 2.3 mL, sterile, case of 50 ea (10 bags of 5ea)
description
96 square wells, U-bottom wells, H-style, capacity 2.2 mL, non-sterile, case of 50 ea (10 bags of 5ea)
description
96 round wells, U-bottom wells, capacity 2.0 mL, non-sterile, case of 50 ea (10 bags of 5ea)
feature
skirt: no, lid: no
feature
skirt: no, lid: no
feature
lid: no, skirt: no
feature
lid: no, skirt: no
manufacturer/tradename
MTC™ Bio D3096-45
manufacturer/tradename
MTC™ Bio D3096-45S
manufacturer/tradename
MTC™ Bio D3096-43
manufacturer/tradename
MTC™ Bio D3096-37
sterility
non-sterile
sterility
sterile
sterility
non-sterile
sterility
non-sterile
material
clear wells, square wells, clear plate, polypropylene , V-bottom wells
material
clear plate, square wells, polypropylene , V-bottom wells, clear wells
material
U-bottom wells, clear wells, clear plate, square wells, polypropylene
material
polypropylene , clear wells, U-bottom wells, round wells, clear plate
size
96 wells
size
96 wells
size
96 wells
size
96 wells
packaging
case of 50 ea (10 bags of 5ea)
packaging
case of 50 ea (10 bags of 5ea)
packaging
case of 50 ea (10 bags of 5ea)
packaging
case of 50 ea (10 bags of 5ea)
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)