Vai a
MTCH680010
MTC™ Bio Halo™ Lightweight Single Channel Pipettor
capacity 0.5-10 μL
Sinonimo/i:
pipette, MTC Bio H6800-10, handheld liquid transfer device, liquid transfer device, mechanical pipettor
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
Informazioni su questo articolo
NACRES:
NB.15
UNSPSC Code:
41121500
Packaging:
pack of 1 ea
Feature:
adjustable, manual, no. of channels 1
Manufacturer/tradename:
MTC™ Bio H6800-10
Prezzi e disponibilità al momento non sono disponibili
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
feature
adjustable, manual, no. of channels 1
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio H6800-10
capacity
0.5-10 μL
General description
As one of the lightest pipettors ever introduced, the Halo™ Pipettor dramatically reduces the risk of repetitive stress injury and the general shoulder, arm, and hand fatigue associated with daily pipetting. Reduced actuation forces andan ergonomic design make Halo™ Pipettors feel balanced and natural in hand. Featuring a four-digit micrometer display for precise volume selection and a click-based adjustment wheel with DriftGuard protection, the Halo™ Pipettor is perfect for applications that demand exceptional accuracy. With a recessed ejector button, and cushioned, shock-absorbing stop, the ergonomics of tip ejection have been re-imagined to relieve stress on the thumb and joints.
Halo™ Single Channel Pipettor, lightweight, 0.1-2.0µL
Features and Benefits
  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips
Legal Information
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
MTCH680010MMTCH68001MMTCH680050
description
capacity 0.5-10 μL
description
capacity 1-100 μL
description
capacity 100-1000 μL
description
capacity 5-50 μL
feature
manual, adjustable, no. of channels 1
feature
manual, adjustable, no. of channels 1
feature
manual, adjustable, no. of channels 1
feature
no. of channels 1, adjustable, manual
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
capacity
0.5-10 μL
capacity
1-100 μL
capacity
100-1000 μL
capacity
5-50 μL
manufacturer/tradename
MTC™ Bio H6800-10
manufacturer/tradename
MTC™ Bio H6800-10M
manufacturer/tradename
MTC™ Bio H6800-1M
manufacturer/tradename
MTC™ Bio H6800-50
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)