Merck
  • A formulation strategy for gamma secretase inhibitor ELND006, a BCS class II compound: development of a nanosuspension formulation with improved oral bioavailability and reduced food effects in dogs.
A formulation strategy for gamma secretase inhibitor ELND006, a BCS class II compound: development of a nanosuspension formulation with improved oral bioavailability and reduced food effects in dogs.
Journal of pharmaceutical sciences (2012-01-04)
Kevin Quinn, Rampurna Prasad Gullapalli, Elaine Merisko-Liversidge, Erich Goldbach, Angelina Wong, Gary G Liversidge, Wherly Hoffman, John-Michael Sauer, John Bullock, George Tonn
ABSTRACT
ELND006 is a novel gamma secretase inhibitor previously under investigation for the oral treatment of Alzheimer's disease. ELND006 shows poor solubility and has moderate to high permeability, suggesting it is a Biopharmaceutics Classification System Class II compound. The poor absolute oral bioavailability of the compound in fasted dogs (F ∼11%) is attributed to poor aqueous solubility. In addition, inhibiting amyloid precursor protein but not Notch cleavage is an important goal for gamma secretase inhibitors; therefore, significant variation in bioavailability resulting from food consumption is a potential liability for this class of compounds. The objective of the present study was to determine if an ELND006 nanocrystalline formulation would offer improved and predictable pharmacokinetics. ELND006 was formulated as a nanosuspension with a mean particle size of less than 200 nm, which was stable in particle size and crystallinity for over 1 year. In addition, ELND006 nanosuspension exhibited rapid dissolution in comparison with reference active pharmaceutical ingredient (API). The in vivo performance of the ELND006 nanosuspension was tested in fed and fasted beagle dogs and compared with a gelatin capsule containing reference API. The results show that nanosizing ELND006 profoundly improved the oral bioavailability and virtually eliminated variation resulting from food intake.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
UCSF924, ≥98% (HPLC)
Sigma-Aldrich
ELND006, ≥98% (HPLC)