Merck
Defining the dopamine transporter proteome by convergent biochemical and in silico analyses.
Genes, brain, and behavior (2006-04-29)
R Maiya, I Ponomarev, K D Linse, R A Harris, R D Mayfield
ABSTRACT
Monoamine transporters play a key role in neuronal signaling by mediating reuptake of neurotransmitters from the synapse. The function of the dopamine transporter (DAT), an important member of this family of transporters, is regulated by multiple signaling mechanisms, which result in altered cell surface trafficking of DAT. Protein-protein interactions are likely critical for this mode of transporter regulation. In this study, we identified proteins associated with DAT by immunoprecipitation (IP) followed by mass spectrometry. We identified 20 proteins with diverse cellular functions that can be classified as trafficking proteins, cytoskeletal proteins, ion channels and extracellular matrix-associated proteins. DAT was found to associate with the voltage-gated potassium channel Kv2.1 and synapsin Ib, a protein involved in regulating neurotransmitter release. An in silico analysis provided evidence for common transcriptional regulation of the DAT proteome genes. In summary, this study identified a network of proteins that are primary candidates for functional regulation of the DAT, an important player in mechanisms of mental disorders and drug addiction.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-Potassium Channel Kv2.1 (DRK1) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder