Merck
  • NLRP12 Promotes Mouse Neutrophil Differentiation through Regulation of Non-canonical NF-κB and MAPKERK1/2 Signaling.
NLRP12 Promotes Mouse Neutrophil Differentiation through Regulation of Non-canonical NF-κB and MAPKERK1/2 Signaling.
International journal of biological sciences (2018-02-28)
Qian Wang, Furao Liu, Meichao Zhang, Pingting Zhou, Ci Xu, Yanyan Li, Lei Bian, Yuanhua Liu, Yuan Yao, Fei Wang, Yong Fang, Dong Li
ABSTRACT
Neutrophils are the most important component of the innate immune system. Mechanistic understanding of the mechanism underlying neutrophil differentiation remains elusive. Using genome-wide RNA-seq, we identified genes whose expression is dramatically up-regulated during neutrophil differentiation. Among them is nucleotide-binding leucine-rich repeat and pyrindomain-containing receptor 12 (NLRP12), which plays a role in immune inflammatory responses. Genetic ablation of NLRP12 suppresses NF-κB inducing kinase (NIK) stabilization, RelB nuclear translocation and neutrophil differentiation in vitro. At a mechanistic level, NLRP12 inhibits the activity of mitogen-activated protein kinases (MAPK)/extracellular signal-regulated kinases (ERK1/2), relieves ERK1/2 suppression of NIK protein levels. Thus, NLRP12 enhances noncanonical NF-κB signaling through inhibition of ERK1/2 signaling, thereby promoting neutrophil differentiation.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-NALP12 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution