Merck
Protein kinase D3 modulates MMP1 and MMP13 expression in human chondrocytes.
PloS one (2018-04-14)
Jonathan Baker, Adrian M D Falconer, David J Wilkinson, G Nicholas Europe-Finner, Gary J Litherland, Andrew D Rowan
ABSTRACT
Many catabolic stimuli, including interleukin-1 (IL-1) in combination with oncostatin M (OSM), promote cartilage breakdown via the induction of collagen-degrading collagenases such as matrix metalloproteinase 1 (MMP1) and MMP13 in human articular chondrocytes. Indeed, joint diseases with an inflammatory component are characterised by excessive extracellular matrix (ECM) catabolism. Importantly, protein kinase C (PKC) signalling has a primary role in cytokine-induced MMP1/13 expression, and is known to regulate cellular functions associated with pathologies involving ECM remodelling. At present, substrates downstream of PKC remain undefined. Herein, we show that both IL-1- and OSM-induced phosphorylation of protein kinase D (PKD) in human chondrocytes is strongly associated with signalling via the atypical PKCι isoform. Consequently, inhibiting PKD activation with a pan-PKD inhibitor significantly reduced the expression of MMP1/13. Specific gene silencing of the PKD isoforms revealed that only PKD3 (PRKD3) depletion mirrored the observed MMP repression, indicative of the pharmacological inhibitor specifically affecting only this isoform. PRKD3 silencing was also shown to reduce serine phosphorylation of signal transducer and activator of transcription 3 (STAT3) as well as phosphorylation of all three mitogen-activated protein kinase groups. This altered signalling following PRKD3 silencing led to a significant reduction in the expression of the activator protein-1 (AP-1) genes FOS and JUN, critical for the induction of many MMPs including MMP1/13. Furthermore, the AP-1 factor activating transcription factor 3 (ATF3) was also reduced concomitant with the observed reduction in MMP13 expression. Taken together, we highlight an important role for PKD3 in the pro-inflammatory signalling that promotes cartilage destruction.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo clone 6C5 anti-gliceraldeide-3-fosfato deidrogenasi, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKD1