Merck
Collagen 1A1 (COL1A1) promotes metastasis of breast cancer and is a potential therapeutic target.
Discovery medicine (2018-06-16)
Jing Liu, Jia-Xin Shen, Hua-Tao Wu, Xiao-Li Li, Xiao-Fen Wen, Cai-Wen Du, Guo-Jun Zhang
ABSTRACT
Extracellular matrix (ECM) is an important component of tumor microenvironment and plays critical roles in cancer development and metastasis, in which collagen is the major structural protein. Collagen type I alpha 1 (COL1A1) is reportedly associated with the development of several human diseases. However, the functions and mechanisms of cellular expression of COL1A1 in breast cancer remain unknown. The purpose of this study is to investigate the cellular expression of COL1A1 in breast cancer cells and patients, and its role in the development and metastasis of breast cancer. The immunofluorescence staining was used to identify the cellular location of COL1A1 in breast cancer cell lines. Real-time PCR was applied to measuring the mRNA levels of COL1A1 and genes of interest. Wound healing and transwell assay were performed to evaluate the effect of COL1A1 on metastasis of breast cancer cells. 97 patients with breast cancer were recruited in this study for evaluating the correlation of COL1A1 with survival and clinicopathological parameters. COL1A1 was expressed in all examined breast cancer cells. Knockdown of COL1A1 inhibited metastasis of breast cancer cells, with a low-level of CXCR4, independent of the epithelial-mesenchymal transition (EMT) process. In patients with breast cancer, cellular expression of COL1A1 was associated with ER/PR expression and metastasis status. The increased COL1A1 level was associated with poor survival, especially in patients with ER+ breast cancer. Patients with a high-level of COL1A1 showed better cisplatin-based chemotherapy response. Cellular expression of COL1A1 could promote breast cancer metastasis. COL1A1 is a new prognostic biomarker and a potential therapeutic target for breast cancer, especially in ER+ patients.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human COL1A1