Merck
  • Systems-wide Analysis of Serine ADP-Ribosylation Reveals Widespread Occurrence and Site-Specific Overlap with Phosphorylation.
Systems-wide Analysis of Serine ADP-Ribosylation Reveals Widespread Occurrence and Site-Specific Overlap with Phosphorylation.
Cell reports (2018-08-30)
Sara C Larsen, Ivo A Hendriks, David Lyon, Lars J Jensen, Michael L Nielsen
ABSTRACT
ADP-ribosylation (ADPr) is a reversible posttranslational modification involved in a range of cellular processes. Here, we report system-wide identification of serine ADPr in human cells upon oxidative stress. High-resolution mass spectrometry and unrestricted data processing confirm that serine residues are the major target of ADPr in HeLa cells. Proteome-wide analysis identifies 3,090 serine ADPr sites, with 97% of acceptor sites modulating more than 2-fold upon oxidative stress, while treatment with the poly (ADP-ribose) polymerase (PARP) inhibitor olaparib abrogates this induction. Serine ADPr predominantly targets nuclear proteins, while structural-predictive analyses reveal that serine ADPr preferentially targets disordered protein regions. The identified ADP-ribosylated serines significantly overlap with known phosphorylated serines, and large-scale phosphoproteomics analysis provides evidence for site-specific crosstalk between serine ADPr and phosphorylation. Collectively, we demonstrate that serine ADPr is a widespread modification and a major nuclear signaling response to oxidative stress, with a regulatory scope comparable to other extensive posttranslational modifications.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
PARG human, recombinant, expressed in Sf21 cells, His tagged, >95% (SDS-PAGE)