Merck
  • Chebulagic acid Chebulinic acid and Gallic acid, the active principles of Triphala, inhibit TNFα induced pro-angiogenic and pro-inflammatory activities in retinal capillary endothelial cells by inhibiting p38, ERK and NFkB phosphorylation.
Chebulagic acid Chebulinic acid and Gallic acid, the active principles of Triphala, inhibit TNFα induced pro-angiogenic and pro-inflammatory activities in retinal capillary endothelial cells by inhibiting p38, ERK and NFkB phosphorylation.
Vascular pharmacology (2018-04-22)
Sivasankar Shanmuganathan, Narayanasamy Angayarkanni
ABSTRACT
Tumor necrosis factor-α (TNFα) a pleiotropic cytokine induces pro-inflammatory and pro-angiogenic changes in conditions such as diabetic retinopathy (DR) and neovascular age related macular degeneration (NV-AMD). Hence, inhibition of TNFα mediated changes can benefit the management of DR and NV-AMD. Triphala, an ayurvedic herbal preparation is known to have immunomodulatry functions. In this study we evaluated the alcoholic extract of triphala (AlE) and its compounds Chebulagic acid (CA), Chebulinic acid (CI) and Gallic acid (GA) for their anti-TNFα activity. TNFα induced pro-inflammatory and pro-angiogenic changes in the retinal-choroid microvascular endothelial cells (RF/6A). Treatment with CA/CI/GA and the whole Triphala extract showed characteristic inhibition of MMP-9, cell proliferation/migration and tube formation as well the expression of IL-6, IL-8 and MCP-1 without affecting cell viability. This was mediated by inhibition of p38, ERK and NFκB phosphorylation. Ex vivo angiogenesis assay using chick chorioallantoic membrane (CAM) model also showed that TNFα-induced angiogenesis and it was inhibited by AlE and its active principles. Further, in silico studies revealed that CA, CI and GA are capable of binding the TNFα-receptor-1 to mediate anti-TNFα activity. This study explains the immunomodulatory function of Triphala, evaluated in the context of retinal and choroid vasculopathies in vitro and ex vivo; which showed that CA, CI and GA can be a potential pharmacological agents in the management of DR and NV-AMD.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Chebulagic acid, analytical standard