Merck
ADIPOR1 is essential for vision and its RPE expression is lost in the Mfrprd6 mouse.
Scientific reports (2018-09-27)
Valentin M Sluch, Angela Banks, Hui Li, Maura A Crowley, Vanessa Davis, Chuanxi Xiang, Junzheng Yang, John T Demirs, Joanna Vrouvlianis, Barrett Leehy, Shawn Hanks, Alexandra M Hyman, Jorge Aranda, Bo Chang, Chad E Bigelow, Dennis S Rice
ABSTRACT
The knockout (KO) of the adiponectin receptor 1 (AdipoR1) gene causes retinal degeneration. Here we report that ADIPOR1 protein is primarily found in the eye and brain with little expression in other tissues. Further analysis of AdipoR1 KO mice revealed that these animals exhibit early visual system abnormalities and are depleted of RHODOPSIN prior to pronounced photoreceptor death. A KO of AdipoR1 post-development either in photoreceptors or the retinal pigment epithelium (RPE) resulted in decreased expression of retinal proteins, establishing a role for ADIPOR1 in supporting vision in adulthood. Subsequent analysis of the Mfrprd6 mouse retina demonstrated that these mice are lacking ADIPOR1 in their RPE layer alone, suggesting that loss of ADIPOR1 drives retinal degeneration in this model. Moreover, we found elevated levels of IRBP in both the AdipoR1 KO and the Mfrprd6 models. The spatial distribution of IRBP was also abnormal. This dysregulation of IRBP hypothesizes a role for ADIPOR1 in retinoid metabolism.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Amido Black Staining Solution 2X, electrophoresis reagent