Merck
  • GC-MS methods for metabolic profiling of microbial fermentation products of dietary polyphenols in human and in vitro intervention studies
GC-MS methods for metabolic profiling of microbial fermentation products of dietary polyphenols in human and in vitro intervention studies
Journal of Chromatography. B, Biomedical Sciences and Applications (2008)
Grun CH, et al.