Merck
Selective mono- and 1,2-difunctionalisation of cyclopentene derivatives via Mg and Cu intermediates.
Chemistry (Weinheim an der Bergstrasse, Germany) (2008-01-31)
Christina Despotopoulou, Richard C Bauer, Arkady Krasovskiy, Peter Mayer, Jeffrey M Stryker, Paul Knochel
ABSTRACT
A single Br/Mg exchange of 1,2-dibromocyclopentene with iPrMgCl LiCl provides the corresponding beta-bromocyclopentenylmagnesium reagent, which can then be reacted with various electrophiles (yields: 65-82 %). In the presence of a secondary alkylmagnesium halide and Li2CuCl4 (2 mol %), these 2-bromoalkenylmagnesium compounds undergo bromine substitution and can then further react with electrophiles to give 1,2-difunctionalised cyclopentenes (63-79 %). The mechanism of this process is discussed.