Merck
  • Proteinase-activated receptors induce interleukin-8 expression by intestinal epithelial cells through ERK/RSK90 activation and histone acetylation.
Proteinase-activated receptors induce interleukin-8 expression by intestinal epithelial cells through ERK/RSK90 activation and histone acetylation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2010-01-13)
Hongying Wang, France Moreau, Christina L Hirota, Wallace K MacNaughton
ABSTRACT
Proteinase-activated receptors (PARs) are involved in both inflammation and tumorigenesis in epithelial cells. Interleukin (IL)-8 is a potent chemoattractant and is also involved in angiogenesis. The molecular mechanism whereby PARs induce epithelial IL-8 expression is not known. In HT-29 colonic epithelial cells, PAR(1) or PAR(2) agonists stimulated the expression of IL-8 through a NF-kappaB-dependent pathway without inducing IkappaB degradation and disassociation of IkappaB from NF-kappaB. Further studies revealed that PAR activation induced the phosphorylation of p65 at Ser-276 in the nucleus, which increased the recruitment of histone acetyltransferase (HAT) p300 to p50. Inhibition of ERK activation completely blocked PAR-induced IL-8 expression, phosphorylation of p65 and HAT activity. We also demonstrated that RSK p90 was the downstream kinase that mediated ERK-induced nuclear p65 phosphorylation. In conclusion, activation of either PAR(1) or PAR(2) stimulated the transcriptional up-regulation of IL-8 in HT-29 colonic epithelial cells through a pathway that involved ERK/RSK p90, NF-kappaB phosphorylation, and HAT activity. These studies provide evidence of a new role for serine proteinases and PARs in the regulation of gene expression in colonic inflammation and tumorigenesis.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo anti-acetil-istone H3, from rabbit
Sigma-Aldrich
Anti-NFκB p65 Antibody, CT, Upstate®, from rabbit