Merck
Adenovirus hexon protein is a potent adjuvant for activation of a cellular immune response.
Journal of virology (2001-12-12)
Valérie Molinier-Frenkel, Renée Lengagne, Florence Gaden, Saw-See Hong, Jeannine Choppin, Hanne Gahery-Ségard, Pierre Boulanger, Jean-Gérard Guillet
ABSTRACT
The capacity of recombinant adenoviruses (rAd) to induce immunization against their transgene products has been well documented. In the present study, we evaluated the vaccinal adjuvant role of rAd independently of its vector function. BALB/c mice received one subcutaneous injection of a mixture of six lipopeptides (LP6) used as a model immunogen, along with AdE1 degrees (10(9) particles), a first-generation rAd empty vector. Although coinjected with a suboptimal dose of lipopeptides, AdE1 degrees significantly improved the effectiveness of the vaccination, even in the absence of booster immunization. In contrast to mice that received LP6 alone or LP6 plus a mock adjuvant, mice injected with AdE1 degrees plus LP6 developed both a polyspecific T-helper type 1 response and an effector CD8 T-cell response specific to at least two class I-restricted epitopes. The helper response was still observed when immunization was performed using LP6 plus a mixture of soluble capsid components released from detergent-disrupted virions. When mice were immunized with LP6 and each individual capsid component, i.e., hexon, penton base, or fiber, the results obtained suggested that hexon protein was responsible for the adjuvant effect exerted by disrupted Ad particles on the helper response to the immunogen. Our results thus have some important implications not only in vaccinology but also for gene therapy using rAd vectors.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
DEAE–Sepharose, Fast Flow