Merck
  • Protein kinase C modulates agonist-sensitive release of Ca2+ from internal stores in HEK293 cells overexpressing the calcium sensing receptor.
Protein kinase C modulates agonist-sensitive release of Ca2+ from internal stores in HEK293 cells overexpressing the calcium sensing receptor.
The Journal of biological chemistry (2004-12-02)
Amos M Sakwe, Lars Rask, Erik Gylfe
ABSTRACT
This study examined the mechanism of Ca2+ entry and the role of protein kinase C (PKC) in Ca2+ signaling induced by activation of the calcium sensing receptor (CaR) in HEK293 cells stably expressing the CaR. We demonstrate that influx of Ca2+ following CaR activation exhibits store-operated characteristics in being associated with Ca2+ store depletion and inhibited by 2-aminoethoxydiphenyl borate. Inhibition of PKC with GF109203X, Go6983, or Go6976 and down-regulation of PKC activity enhanced the release of Ca2+ from internal stores in response to the polyvalent cationic CaR agonist neomycin, whereas activation of PKC with acute 12-O-tetradecanoylphorbol-13-acetate treatment decreased the release. In contrast, overexpression of wild type PKC-alpha or -epsilon augmented the neomycin-induced release of Ca2+ from internal stores, whereas dominant negative PKC-epsilon strongly decreased the release, but dominant negative PKC-alpha had little effect. Prolonged treatment of cells with 12-O-tetradecanoylphorbol-13-acetate effectively down-regulated immunoreactive PKC-alpha but had little effect on the expression of PKC-epsilon. Together these results indicate that diacylglycerol-responsive PKC isoforms differentially influence CaR agonist-induced release of Ca2+ from internal stores. The fundamentally different results obtained when overexpressing or functionally down-regulating specific PKC isoforms as compared with pharmacological manipulation of PKC activity indicate the need for caution when interpreting data obtained with the latter approach.