Merck
  • The protective effects of chronic intermittent hypobaric hypoxia pretreatment against collagen-induced arthritis in rats
The protective effects of chronic intermittent hypobaric hypoxia pretreatment against collagen-induced arthritis in rats
Journal of Inflammation (2015)
Shi M, et al.