Merck
  • Modification and comparison of three Gracilaria spp. agarose with methylation for promotion of its gelling properties
Modification and comparison of three Gracilaria spp. agarose with methylation for promotion of its gelling properties
Chemistry Central Journal (2017)
Gu Y, et al.