Merck
  • Combination of Microwave-Assisted Girard Derivatization with Ionic Liquid Matrix for Sensitive MALDI-TOF MS Analysis of Human Serum N-Glycans
Combination of Microwave-Assisted Girard Derivatization with Ionic Liquid Matrix for Sensitive MALDI-TOF MS Analysis of Human Serum N-Glycans
Journal of Analytical Methods in Chemistry (2018)
Le HT, et al.