Merck
  • Development and validation of liquid chromatography and capillary electrophoresis methods for acarbose determination in pharmaceutical tablets.
Development and validation of liquid chromatography and capillary electrophoresis methods for acarbose determination in pharmaceutical tablets.
Journal of Pharmaceutical and Biomedical Analysis (1998)
Cherkaoui S, et al.