Merck
  • Effect of milking frequency and α-tocopherol plus selenium supplementation on sheep milk lipid composition and oxidative stability.
Effect of milking frequency and α-tocopherol plus selenium supplementation on sheep milk lipid composition and oxidative stability.
Journal of dairy science (2019-02-18)
E Pulido, M Fernández, N Prieto, R L Baldwin, S Andrés, S López, F J Giráldez
ABSTRACT
The aim of this research was to study the effect of milking frequency [once-daily milking (ODM) vs. twice-daily milking (TDM)] and antioxidant (AOX) supplementation on fatty acid (FA) profile and oxidative stability in sheep milk. Sixteen Assaf ewes were used; 8 did not receive any vitamin-mineral supplement (control), and the other 8 received an oral dose of 1,000 IU of α-tocopherol and 0.4 mg of Se daily. The experiment consisted of 2 consecutive periods; the first was 3 wk with TDM of both mammary glands. The second period was 8 wk and consisted of ODM of one mammary gland and TDM of the other gland. All ewes were fed ad libitum the same total mixed ration from lambing and throughout the experiment. There were no differences in plasma or milk Se concentrations between control and AOX ewes. However, plasma and milk α-tocopherol concentrations and AOX capacity were increased in ewes receiving the AOX supplement. Milk FA profile was practically unaffected after 21 d of AOX supplementation. However, after 77 d, AOX supplementation increased the relative percentage of C16:0 and cis-9 C18:1 and reduced the proportions of some saturated FA with less than 16 carbons and cis-9 C12:1. Antioxidant supplementation had no effect on the proportions of conjugated linoleic acid or total polyunsaturated FA (PUFA) but decreased the proportion of trans-7,cis-9 C18:2 and increased that of n-6 C20:3. Once-daily milking did not affect α-tocopherol, Se, or fat resistance to oxidation in milk. Total monounsaturated FA, cis-9 C16:1, and several cis and trans isomers of C18:1 were increased and total saturated FA were decreased in milk from ODM glands. Compared with TDM, ODM increased the proportions of cis-9,cis-12 C18:2 and several isomers of C18:2 and reduced those of cis-9,cis-12,cis-15 C18:3 and some PUFA of 20 and 22 carbons, but total proportion of PUFA was unaffected. Once-daily milking and AOX supplementation modified milk FA profile, but the effects of ODM could be considered of little biological relevance for consumer health. Supplementing ewes with α-tocopherol plus Se could be considered an effective strategy to improve plasma AOX status and reduce milk fat oxidation without substantial changes in the milk FA profile.