Merck
Liver tumor promotion by the cyanobacterial cyclic peptide toxin microcystin-LR.
Journal of cancer research and clinical oncology (1992-01-01)
R Nishiwaki-Matsushima, T Ohta, S Nishiwaki, M Suganuma, K Kohyama, T Ishikawa, W W Carmichael, H Fujiki
ABSTRACT
Certain waterblooms of toxic cyanobacteria (blue-green algae) are a health threat because of their production of toxic peptides, termed microcystins, which cause liver damage in wild and domesticated animals. The most widely studied microcystin is microcystin-LR, a heptapeptide containing the two L-amino acids, leucine and arginine. The inhibition of protein phosphatase type 1 and type 2A activities by microcystin-LR is similar to that of the known protein phosphatase inhibitor and tumor promoter okadaic acid. We show in this report that microcystin-LR, applied below the acute toxicity level, dose-dependently increases the number and percentage area of positive foci for the placental form of glutathione S-transferase in rat liver, which was initiated with diethylnitrosamine. The result was obtained independently through two animal experiments. This observation indicates that microcystin-LR is a new liver tumor promoter mediated through inhibition of protein phosphatase type 1 and type 2A activities. This provides further evidence that the okadaic acid pathway is a general mechanism of tumor promotion in various organs, such as mouse skin, rat glandular stomach and rat liver.