Merck
  • High-performance liquid chromatographic analysis of biogenic amines in pharmaceutical products containing Citrus aurantium
High-performance liquid chromatographic analysis of biogenic amines in pharmaceutical products containing Citrus aurantium
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment (2013)
Viana C, et al.