Merck
  • Normal endothelial but impaired arterial development in MAP-Kinase activated protein kinase 2 (MK2) deficient mice
Normal endothelial but impaired arterial development in MAP-Kinase activated protein kinase 2 (MK2) deficient mice
Vascular Cell (2016)
Napp LC, et al.