Merck
GRP94 Is Involved in the Lipid Phenotype of Brain Metastatic Cells.
International journal of molecular sciences (2019-08-10)
Naiara Santana-Codina, Anna Marcé-Grau, Laia Muixí, Claudia Nieva, Mónica Marro, David Sebastián, Juan Pablo Muñoz, Antonio Zorzano, Angels Sierra
ABSTRACT
Metabolic adaptation may happen in response to the pressure exerted by the microenvironment and is a key step in survival of metastatic cells. Brain metastasis occurs as a consequence of the systemic dissemination of tumor cells, a fact that correlates with poor prognosis and high morbidity due to the difficulty in identifying biomarkers that allow a more targeted therapy. Previously, we performed transcriptomic analysis of human breast cancer patient samples and evaluated the differential expression of genes in brain metastasis (BrM) compared to lung, bone and liver metastasis. Our network approach identified upregulation of glucose-regulated protein 94 (GRP94) as well as proteins related to synthesis of fatty acids (FA) in BrM. Here we report that BrM cells show an increase in FA content and decreased saturation with regard to parental cells measured by Raman spectroscopy that differentiate BrM from other metastases. Moreover, BrM cells exerted a high ability to oxidize FA and compensate hypoglycemic stress due to an overexpression of proteins involved in FA synthesis and degradation (SREBP-1, LXRα, ACOT7). GRP94 ablation restored glucose dependence, down-regulated ACOT7 and SREBP-1 and decreased tumorigenicity in vivo. In conclusion, GRP94 is required for the metabolic stress survival of BrM cells, and it might act as a modulator of lipid metabolism to favor BrM progression.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human HSP90B1