Merck
Cell Death Induced by Cationic Amphiphilic Drugs Depends on Lysosomal Ca2+ Release and Cyclic AMP.
Molecular cancer therapeutics (2019-07-10)
Atul Anand, Bin Liu, Jano Dicroce Giacobini, Kenji Maeda, Mikkel Rohde, Marja Jäättelä
ABSTRACT
Repurposing cationic amphiphilic drugs (CAD) for cancer treatment is emerging as an attractive means to enhance the efficacy of chemotherapy. Many commonly used CADs, including several cation amphiphilic antihistamines and antidepressants, induce cancer-specific, lysosome-dependent cell death and sensitize cancer cells to chemotherapy. CAD-induced inhibition of lysosomal acid sphingomyelinase is necessary, but not sufficient, for the subsequent lysosomal membrane permeabilization and cell death, while other pathways regulating this cell death pathway are largely unknown. Prompted by significant changes in the expression of genes involved in Ca2+ and cyclic AMP (cAMP) signaling pathways in CAD-resistant MCF7 breast cancer cells, we identified here an early lysosomal Ca2+ release through P2X purinergic receptor 4 (P2RX4) and subsequent Ca2+- and adenylyl cyclase 1 (ADCY1)-dependent synthesis of cAMP as a signaling route mediating CAD-induced lysosomal membrane permeabilization and cell death. Importantly, pharmacologic and genetic means to increase cellular cAMP levels either by activating cAMP-inducing G-protein-coupled receptors (GPR3 or β2 adrenergic receptor) or ADCY1, or by inhibiting cAMP-reducing guanine nucleotide-binding protein G(i) subunit α2, C-X-C motif chemokine receptor type 4, or cAMP phosphodiesterases, sensitized cancer cells to CADs. These data reveal a previously unrecognized lysosomal P2RX4- and ADCY1-dependent signaling cascade as a pathway essential for CAD-induced lysosome-dependent cell death and encourage further investigations to find the most potent combinations of CADs and cAMP-inducing drugs for cancer therapy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human ADCY1