Merck
  • In situ surface modification on dental composite resin using 2-methacryloyloxyethyl phosphorylcholine polymer for controlling plaque formation.
In situ surface modification on dental composite resin using 2-methacryloyloxyethyl phosphorylcholine polymer for controlling plaque formation.
Materials science & engineering. C, Materials for biological applications (2019-09-11)
Jun Koyama, Kyoko Fukazawa, Kazuhiko Ishihara, Yoshiyuki Mori
ABSTRACT
Composite resins (CRs) are widely used as dental restorative materials for caries treatment. They cause problems of secondary caries since Streptococcus mutans stays in the dental plaque, which the surface exists and produces acidic compounds during metabolism. The dental plaque depositions are induced by the protein adsorption on the surface. Therefore, suppression of protein adsorption on the surface of the CRs is important for inhibiting the formation of plaque and secondary caries. In this study we developed a surface treatment to provide an antibiofouling nature to the CRs by chemical reaction with 2-methacryloyloxyethyl phosphorylcholine (MPC) polymers in the oral cavity during dental treatment. To carry out the photochemical reaction on the remaining polymerizable groups of CRs, we synthesized the MPC polymer with a polymerizable group in the side chain. The MPC polymer could bind on the surfaces of the CRs chemically under dental treatment procedures. The treated surface showed significant resistance to oral protein adsorption and bacterial adhesion even when the surface was brushed with a toothbrush. Thus, we concluded that the photochemical reaction of the MPC polymer with the CRs in the oral cavity was good for making an antibiofouling surface and preventing secondary caries.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
2-Aminoethyl methacrylate hydrochloride, contains ~500 ppm phenothiazine as stabilizer, 90%
Sigma-Aldrich
Pentafluorophenyl methacrylate, contains MEHQ as inhibitor, 95%