Merck
Mitochondrial morphology and dynamics in hepatocytes from normal and ethanol-fed rats.
Pflugers Archiv : European journal of physiology (2012-04-25)
Sudipto Das, Nora Hajnóczky, Anil Noronha Antony, György Csordás, Lawrence D Gaspers, Dahn L Clemens, Jan B Hoek, György Hajnóczky
ABSTRACT
Mitochondrial structure and function are central to cell physiology and are mutually interdependent. Mitochondria represent a primary target of the alcohol-induced tissue injury, particularly in the liver, where the metabolic effects of ethanol are predominant. However, the effect of ethanol on hepatic mitochondrial morphology and dynamics remain to be established. In the present work, we employed the organelle-targeted photoactivatable fluorescent protein technology and electron microscopy to study hepatic mitochondrial structure and dynamics. Hepatocytes in perfused liver as well as in primary cultures showed mostly discrete globular or short tubular mitochondria. The mitochondria showed few fusion events and little movement activity. By contrast, human hepatoma (HepG2)-derived VL-17A cells, expressing the major hepatic ethanol metabolizing enzymes, alcohol dehydrogenase and cytochrome P450 2E1, have elongated and interconnected mitochondria showing matrix continuity and many fusion events. Hepatocytes isolated from chronically ethanol-fed rats showed some increase in mitochondrial volume and exhibited a substantial suppression of mitochondrial dynamics. In VL-17A cells, prolonged ethanol exposure also caused decreased mitochondrial continuity and dynamics. Collectively, these results indicate that mitochondria in normal hepatocytes show relatively slow dynamics, which is very sensitive to suppression by ethanol exposure.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo anti-GRK 4- 6, clone A16/17, clone A16/17, Upstate®, from mouse