Merck
Rapid parallel flow cytometry assays of active GTPases using effector beads.
Analytical biochemistry (2013-08-10)
Tione Buranda, Soumik BasuRay, Scarlett Swanson, Jacob Agola, Virginie Bondu, Angela Wandinger-Ness
ABSTRACT
We describe a rapid assay for measuring the cellular activity of small guanine triphosphatases (GTPases) in response to a specific stimulus. Effector-functionalized beads are used to quantify in parallel multiple GTP-bound GTPases in the same cell lysate by flow cytometry. In a biologically relevant example, five different Ras family GTPases are shown for the first time to be involved in a concerted signaling cascade downstream of receptor ligation by Sin Nombre hantavirus.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo anti-Rac1, clone 23A8, clone 23A8, Upstate®, from mouse
Sigma-Aldrich
Anticorpo anti-Cdc42, from rabbit