Merck
  • Discovery of BPR1K871, a quinazoline based, multi-kinase inhibitor for the treatment of AML and solid tumors: Rational design, synthesis, in vitro and in vivo evaluation.
Discovery of BPR1K871, a quinazoline based, multi-kinase inhibitor for the treatment of AML and solid tumors: Rational design, synthesis, in vitro and in vivo evaluation.
Oncotarget (2016-11-20)
Yung Chang Hsu, Mohane Selvaraj Coumar, Wen-Chieh Wang, Hui-Yi Shiao, Yi-Yu Ke, Wen-Hsing Lin, Ching-Chuan Kuo, Chun-Wei Chang, Fu-Ming Kuo, Pei-Yi Chen, Sing-Yi Wang, An-Siou Li, Chun-Hwa Chen, Po-Chu Kuo, Ching-Ping Chen, Ming-Hsine Wu, Chen-Lung Huang, Kuei-Jung Yen, Yun-I Chang, John T-A Hsu, Chiung-Tong Chen, Teng-Kuang Yeh, Jen-Shin Song, Chuan Shih, Hsing-Pang Hsieh
ABSTRACT
The design and synthesis of a quinazoline-based, multi-kinase inhibitor for the treatment of acute myeloid leukemia (AML) and other malignancies is reported. Based on the previously reported furanopyrimidine 3, quinazoline core containing lead 4 was synthesized and found to impart dual FLT3/AURKA inhibition (IC50 = 127/5 nM), as well as improved physicochemical properties. A detailed structure-activity relationship study of the lead 4 allowed FLT3 and AURKA inhibition to be finely tuned, resulting in AURKA selective (5 and 7; 100-fold selective over FLT3), FLT3 selective (13; 30-fold selective over AURKA) and dual FLT3/AURKA selective (BPR1K871; IC50 = 19/22 nM) agents. BPR1K871 showed potent anti-proliferative activities in MOLM-13 and MV4-11 AML cells (EC50 ~ 5 nM). Moreover, kinase profiling and cell-line profiling revealed BPR1K871 to be a potential multi-kinase inhibitor. Functional studies using western blot and DNA content analysis in MV4-11 and HCT-116 cell lines revealed FLT3 and AURKA/B target modulation inside the cells. In vivo efficacy in AML xenograft models (MOLM-13 and MV4-11), as well as in solid tumor models (COLO205 and Mia-PaCa2), led to the selection of BPR1K871 as a preclinical development candidate for anti-cancer therapy. Further detailed studies could help to investigate the full potential of BPR1K871 as a multi-kinase inhibitor.