Merck
  • Liquid Chromatographic Separation and Thermodynamic Investigation of Ethyl Nipecotate Enantiomers on Immobilized Amylose-Based Chiral Stationary Phase.
Liquid Chromatographic Separation and Thermodynamic Investigation of Ethyl Nipecotate Enantiomers on Immobilized Amylose-Based Chiral Stationary Phase.
Journal of chromatographic science (2019-09-11)
Vipul P Rane, Vinod K Ahirrao, Kiran R Patil, Arjun B Jadhav, Kiran B More, Ravindra D Yeole
ABSTRACT
Ethyl nipecotate enantiomers are widely used as chiral building blocks in the synthesis of drug substances. An efficient and economic chiral high-performance liquid chromatographic method for determination of enantiomeric purity of ethyl nipecotate is developed and validated. Chiral separation was achieved on immobilized amylose-based stationary phase using a mixture of n-hexane: ethanol: diethylamine (80:20:0.1, v/v/v) as mobile phase. Resolution between R-(-)-ethyl nipecotate (REN) and S-(+)-ethyl nipecotate (SEN) peaks was found to be 3.59. The detector response of SEN exhibited an excellent linearity over the concentration range of 4.5-120 μg mL-1. The limit of detection and limit of quantification for SEN were 0.016 and 0.045 μg and REN were 0.015 and 0.043 μg, respectively. The influence of column oven temperatures on chiral retention and separation was studied in the range of 25°C to 50°C; the thermodynamic parameters ΔH°, ΔS° and ΔG° were evaluated from van't Hoff plots and used to explain the strength of interaction between analyte and stationary phase.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Ethyl nipecotate, 96%