Merck
  • Dielectric Relaxation Behavior of Silver Nanoparticles and Graphene Oxide Embedded Poly(vinyl alcohol) Nanocomposite Film: An Effect of Ionic Liquid and Temperature.
Dielectric Relaxation Behavior of Silver Nanoparticles and Graphene Oxide Embedded Poly(vinyl alcohol) Nanocomposite Film: An Effect of Ionic Liquid and Temperature.
Polymers (2020-02-13)
Ganeswar Sahu, Mamata Das, Mithilesh Yadav, Bibhu Prasad Sahoo, Jasaswini Tripathy
ABSTRACT
This paper presents the dielectric characteristics of nanocomposite films of poly(vinyl alcohol) (PVA) embedded with silver (Ag) nanoparticles and graphene oxide(GO). The nanocomposite films were fabricated by using the solvent casting approach. The morphological analysis was carried out through scanning electron microscopy (SEM) and transmission electron microscopy (TEM). The dielectric relaxation behavior of nanocomposite films was analyzed in the frequency range of 101 to 106 Hz, by varying GO loading. The temperature effect was investigated over the temperature range of 40 to 150 °C. The effect of ionic liquid (IL) was also explored by comparing the dielectric behavior of films fabricated without using ionic liquid. The conductive filler loading variation showed a significant effect on dielectric permittivity(ε'), complex impedance (Z*) and electric conductivity (σac). The obtained results revealed that the dielectric permittivity (ε') increased by incorporating Ag nanoparticles and increasing GO loading in PVA matrix. An incremental trend in dielectric permittivity was observed on increasing the temperature, which is attributed to tunneling and hopping mechanism. With an increase in nanofiller loading, the real part of impedance (Z') and imaginary part of impedance (Z'') were found to decrease. Further, the semicircular nature of Nyquist plot indicated the decrease in bulk resistivity on increasing GO loading, temperature and incorporating ionic liquid. On the basis of above findings, the obtained GO-Ag-PVA nanocomposite films can find promising applications in charge storage devices.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1-Allyl-3-methylimidazolium chloride, ≥97.0% (HPLC)