Merck
  • A new synthesis of well-dispersed, core-shell Ag@SiO2 mesoporous nanoparticles using amino acids and sugars.
A new synthesis of well-dispersed, core-shell Ag@SiO2 mesoporous nanoparticles using amino acids and sugars.
Journal of materials chemistry. B (2013-05-21)
Philippe Saint-Cricq, Junzheng Wang, Ayae Sugawara-Narutaki, Atsushi Shimojima, Tatsuya Okubo
ABSTRACT
Mesoporous silica-coated silver nanoparticles (Ag@MSN) were prepared by a two step synthesis in an environment friendly way. Arginine and glucose were used as a catalyst for the formation of silica and as a reducing agent for silver ions, respectively, in the presence of a cationic surfactant. Ag@MSNs having single Ag nanoparticles as cores were obtained with good selectivity. They showed excellent dispersibility in ethanol, forming a uniform coating on a glass substrate. Their antibacterial activity against E. coli was demonstrated.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Esametildisilossano, ≥98%