Merck
  • Novel high-throughput and widely-targeted liquid chromatography-time of flight mass spectrometry method for d-amino acids in foods
Novel high-throughput and widely-targeted liquid chromatography-time of flight mass spectrometry method for d-amino acids in foods
Journal of Bioscience and Bioengineering (2017)
Konya Y, et al.