Merck
SIKIAT1/miR-96/FOXA1 axis regulates sepsis-induced kidney injury through induction of apoptosis.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2020-04-29)
Shijun Lu, Haibin Wu, Jianhua Xu, Zhaofeng He, Haiyan Li, Chao Ning
ABSTRACT
Nowadays, sepsis-induced acute kidney injury (AKI) has gradually become a global problem for its high incidence and increasing mortality. Previous study has reported lncRNA ENST00000452391.1 in sepsis patients. However, its potential biological function and downstream molecular mechanism are still mysterious. METHODS AND RESULTS: Our study found that it was upregulated in sepsis-induced AKI patients, so it was identified as "sepsis-induced kidney injury associated transcript 1 (SIKIAT1)". We used lipopolysaccharide (LPS) stimulated HK-2 cells as an in vitro model to demonstrated that SIKIAT1 acts as a ceRNA for miR-96-3p to enhance FOXA1 expression and promote HK-2 cell apoptosis. Therefore, it could be a potential biomarker and therapeutic target for sepsis-induced AKI in the development of disease.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human FOXA1