Merck
  • Kinetic study of oxytetracycline and chlortetracycline inhibition in the anaerobic digestion of dairy manure.
Kinetic study of oxytetracycline and chlortetracycline inhibition in the anaerobic digestion of dairy manure.
Bioresource technology (2020-07-20)
Fetra J Andriamanohiarisoamanana, Ikko Ihara, Gen Yoshida, Kazutaka Umetsu
ABSTRACT
This study was aimed to investigate the mechanisms of oxytetracycline (OTC) and chlortetracycline (CTC) inhibition in anaerobic digestion (AD) using four kinetic models. The results showed that the inhibition rate of OTC was faster than CTC at OTC and CTC between 0.04 and 1.28 g/L. Hydrolysis rate constant was linearly and positively correlated with OTC and increased from 0.172 to 0.193 d-1, 0.164 to 0.179 d-1 and 0.251 to 0.285 d-1 using first-order kinetic, Fitzhugh and Cone models, respectively, while the maximum specific methane production rate was linearly and negatively correlated with CTC and decreased from 0.028 to 0.016 L/gVS. Cone model was found to give the most satisfactory fitting results followed in descending order by first-order kinetic, Fitzhugh and modified Gompertz models. The kinetic modeling of methane yield helped explain the mechanism of OTC and CTC inhibition in the AD process and provided essential information for further process improvement.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Millipore
Chlortetracycline, suitable for microbiology