Merck
IPEC-1 variable immune response to different serovars of Salmonella enterica subsp. enterica.
Veterinary immunology and immunopathology (2019-12-17)
Mustapha Berri, Daniel Hogan, Georges Saade, Sylvie Roche, Philippe Velge, Isabelle Virlogeux-Payant, François Meurens
ABSTRACT
Salmonella is a genus of Gram-negative bacteria in the Enterobacteriaceae family causing various illnesses. The ability of the different serovars of Salmonella enterica subsp. enterica to infect a host and to induce pathology relies in part on their cellular and molecular interactions with the intestinal epithelium. In the current study, an in vitro approach using non-polarized or polarized IPEC-1 porcine intestinal epithelial cells were used in order to assess the relation between adhesion, invasion, and induction of the immune response as a function of the serotype of Salmonella. Five serovars, Choleraesuis (host-adapted), Typhimurium (ubiquitous), Typhisuis (host-restricted), which are relevant for pig infection, and Dublin and Gallinarum, which are host-restricted or host-adapted, were studied. A strong variation was observed in the percentages of adhesion and invasion amongst the S. enterica serovars used to interact with the non-polarized and polarized cells. Subsequently, differences were identified between serovars in terms of immune response induced. Serovars Typhimurium and Typhisuis induced a strong innate immune response four and half hours after the beginning of cell stimulation while Choleraesuis, Gallinarum, and Dublin did not. A strong inflammatory response could limit the spread of the porcine serovars to the gut while, with a weak response, bacteria may not be constrained by the immune response enabling severe systemic diseases. Different repertoires of adhesion factors and of secreted protein effectors between Salmonella serovars interacting with IPEC-1 cells probably explains the differences in their early pathogenic behaviours.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Millipore
Bilirubin, Principal pigment of bile and one of the major end products of hemoglobin decomposition.