Merck
miR-300/FA2H affects gastric cancer cell proliferation and apoptosis.
Open medicine (Warsaw, Poland) (2020-12-22)
Bo Hong, Jie Li, Chunxiao Huang, Tao Huang, Mengpei Zhang, Lijiang Huang
ABSTRACT
MicroRNA (miR/miRNA) expression disorders play a crucial role in the development of gastric cancer (GC). Increasing evidence has indicated that miRNAs participate in the process of numerous cancers. Previous research has demonstrated that miR-300 acts as a cancer-promoting factor or tumor suppressor in a number of tumors. However, to the best of our knowledge, the effects of miR-300 on GC cells remain largely unknown. The present study investigated the effects of miR-300 on GC cells and analyzed its molecular mechanism. First, reverse transcription-quantitative polymerase chain reaction showed that miR-300 expression was increased in GC tissues and cell lines, with the highest expression observed in human gastric cancer cell line AGS. Subsequent results indicated that fatty acid 2-hydroxylase (FA2H) was a target of miR-300. FA2H-plasmid inhibited AGS cell proliferation and induced apoptosis. Finally, miR-300 inhibitor reduced cell proliferation and induced apoptosis, whereby these effects were reversed by FA2H-small interfering RNA. Therefore, the data demonstrated that miR-300/FA2H might be a new potential biomarker and therapeutic target for GC treatment.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human FA2H