Merck
  • Interleukin-5 promotes ATP-binding cassette transporter A1 expression through miR-211/JAK2/STAT3 pathways in THP-1-dervied macrophages.
Interleukin-5 promotes ATP-binding cassette transporter A1 expression through miR-211/JAK2/STAT3 pathways in THP-1-dervied macrophages.
Acta biochimica et biophysica Sinica (2020-08-14)
Kong Chen, Zhenwang Zhao, Gang Wang, Jin Zou, Xiaohua Yu, Dawei Zhang, Gaofeng Zeng, Chaoke Tang
ABSTRACT
Interleukin-5 (IL-5) is manifested as its involvement in the process of atherosclerosis, but the mechanism is still unknown. In this study, we explored the effect of IL-5 on lipid metabolism and its underlying mechanisms in THP-1-derived macrophages. The quantitative polymerase chain reaction (qPCR) and western blot analysis results showed that IL-5 significantly up-regulated ATP-binding cassette transporter A1 (ABCA1) expression in a dose-dependent and time-dependent manner. [3H]-labeled cholesterol was used to assess the levels of cholesterol efflux, and the results showed that IL-5 increased ABCA1-mediated cholesterol efflux. A high-performance liquid chromatography assay indicated that cellular cholesterol content was decreased by IL-5 treatment in THP-1-derived macrophages. The selective inhibitor and small interfering RNA were used to block the Janus kinase (JAK)/signal transducer and activator of transcription 3 (STAT3) pathway. The results of the qPCR and western blot analysis showed that IL-5 activated JAK2/STAT3 pathway to up-regulate ABCA1 expression. Meanwhile, IL-5 reduced the expression level of miR-211. Furthermore, we found that JAK2 is a target gene of miR-211 and miR-211 mimic inhibited the expression of JAK2 and reduced the levels of p-STAT3 and ABCA1 as revealed by luciferase reporter assay, qPCR and western blot analysis. In summary, these findings indicated that IL-5 promotes ABCA1 expression and cholesterol efflux through the miR-211/JAK2/STAT3 signaling pathway in THP-1-derived macrophages.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3
Sigma-Aldrich
MISSION® esiRNA, targeting human JAK2