Merck
  • Diterpenoids from Gutierrezia sarothrae and G. microcephala: Chemical diversity, chemophenetics and implications to toxicity in grazing livestock.
Diterpenoids from Gutierrezia sarothrae and G. microcephala: Chemical diversity, chemophenetics and implications to toxicity in grazing livestock.
Phytochemistry (2020-09-06)
Dale R Gardner, Daniel Cook, Scott W Larsen, Clinton A Stonecipher, Robert Johnson
ABSTRACT
Broom snakeweed (Gutierrezia sarothrae) and threadleaf snakeweed (G. microcephala) are suffrutescent plants found in many parts of western US rangelands and are possibly toxic to grazing livestock. The toxic components are not known, but it has been suggested that the diterpene acids may be both toxic and abortifacient. One hundred sixty-two samples of snakeweed were collected from 55 locations in Colorado, Oklahoma, New Mexico, Texas, Wyoming and Utah and were taxonomically classified. Samples were analyzed by GC-MS in a chemophenetic analysis and grouped into individual chemotypes based on diterpene acid content. The GC-MS profiles were found to be diverse showing at least eight different chemotypes. From each of the chemotypes the major diterpene acids were isolated and characterized by IR, MS and NMR spectroscopy. Twenty-one diterpenoids were identified and found to be a mix of furano, lactone, di-acid and esters of labdane, ent-labdane and chlerodane acids and alcohols. Only four of the 21 compounds isolated had been previously reported as occurring in G. sarathorea or G. microcephala, while another eight compounds were previously reported from other Gutierrezia or related species. Nine of the isolated diterpenoids have not been previously reported and their structure elucidation is reported.