Merck
Targeting the RhoGEF βPIX/COOL-1 in Glioblastoma: Proof of Concept Studies.
Cancers (2020-12-02)
Kate Connor, David W Murray, Monika A Jarzabek, Nhan L Tran, Kieron White, Patrick Dicker, Kieron J Sweeney, Philip J O'Halloran, Brian MacCarthy, Liam P Shiels, Francesca Lodi, Diether Lambrechts, Jann N Sarkaria, Raymond M Schiffelers, Marc Symons, Annette T Byrne
ABSTRACT
Glioblastoma (GBM), a highly invasive and vascular malignancy is shown to rapidly develop resistance and evolve to a more invasive phenotype following bevacizumab (Bev) therapy. Rho Guanine Nucleotide Exchange Factor proteins (RhoGEFs) are mediators of key components in Bev resistance pathways, GBM and Bev-induced invasion. To identify GEFs with enhanced mRNA expression in the leading edge of GBM tumours, a cohort of GEFs was assessed using a clinical dataset. The GEF βPix/COOL-1 was identified, and the functional effect of gene depletion assessed using 3D-boyden chamber, proliferation, and colony formation assays in GBM cells. Anti-angiogenic effects were assessed in endothelial cells using tube formation and wound healing assays. In vivo effects of βPix/COOL-1-siRNA delivered via RGD-Nanoparticle in combination with Bev was studied in an invasive model of GBM. We found that siRNA-mediated knockdown of βPix/COOL-1 in vitro decreased cell invasion, proliferation and increased apoptosis in GBM cell lines. Moreover βPix/COOL-1 mediated endothelial cell migration in vitro. Mice treated with βPix/COOL-1 siRNA-loaded RGD-Nanoparticle and Bev demonstrated a trend towards improved median survival compared with Bev monotherapy. Our hypothesis generating study suggests that the RhoGEF βPix/COOL-1 may represent a target of vulnerability in GBM, in particular to improve Bev efficacy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Accutase®, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anticorpo anti-α-tubulina, monoclonale murino, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human ARHGEF7