Merck
  • Ablation of the Stimulatory G Protein α-Subunit in Renal Proximal Tubules Leads to Parathyroid Hormone-Resistance With Increased Renal Cyp24a1 mRNA Abundance and Reduced Serum 1,25-Dihydroxyvitamin D.
Ablation of the Stimulatory G Protein α-Subunit in Renal Proximal Tubules Leads to Parathyroid Hormone-Resistance With Increased Renal Cyp24a1 mRNA Abundance and Reduced Serum 1,25-Dihydroxyvitamin D.
Endocrinology (2015-12-17)
Yan Zhu, Qing He, Cumhur Aydin, Isabelle Rubera, Michel Tauc, Min Chen, Lee S Weinstein, Vladimir Marshansky, Harald Jüppner, Murat Bastepe
ABSTRACT
PTH regulates serum calcium, phosphate, and 1,25-dihydroxyvitamin D (1,25(OH)2D) levels by acting on bone and kidney. In renal proximal tubules (PTs), PTH inhibits reabsorption of phosphate and stimulates the synthesis of 1,25(OH)2D. The PTH receptor couples to multiple G proteins. We here ablated the α-subunit of the stimulatory G protein (Gsα) in mouse PTs by using Cre recombinase driven by the promoter of type-2 sodium-glucose cotransporter (Gsα(Sglt2KO) mice). Gsα(Sglt2KO) mice were normophosphatemic but displayed, relative to controls, hypocalcemia (1.19 ±0.01 vs 1.23 ±0.01 mmol/L; P < .05), reduced serum 1,25(OH)2D (59.3 ±7.0 vs 102.5 ±12.2 pmol/L; P < .05), and elevated serum PTH (834 ±133 vs 438 ±59 pg/mL; P < .05). PTH-induced elevation in urinary cAMP excretion was blunted in Gsα(Sglt2KO) mice (2- vs 4-fold over baseline in controls; P < .05). Relative to baseline in controls, PTH-induced reduction in serum phosphate tended to be blunted in Gsα(Sglt2KO) mice (-0.39 ±0.33 vs -1.34 ±0.36 mg/dL; P = .07). Gsα(Sglt2KO) mice showed elevated renal vitamin D 24-hydroxylase and bone fibroblast growth factor-23 (FGF23) mRNA abundance (∼3.4- and ∼11-fold over controls, respectively; P < .05) and tended to have elevated serum FGF23 (829 ±76 vs 632 ±60 pg/mL in controls; P = .07). Heterozygous mice having constitutive ablation of the maternal Gsα allele (E1(m-/+)) (model of pseudohypoparathyroidism type-Ia), in which Gsα levels in PT are reduced, also exhibited elevated serum FGF23 (474 ±20 vs 374 ±27 pg/mL in controls; P < .05). Our findings indicate that Gsα is required in PTs for suppressing renal vitamin D 24-hydroxylase mRNA levels and for maintaining normal serum 1,25(OH)2D.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo anti-Gsα, serum, Upstate®