Merck
Benzo[b]thiophene-based histone deacetylase inhibitors.
Bioorganic & medicinal chemistry letters (2007-06-20)
David J Witter, Sandro Belvedere, Liqiang Chen, J Paul Secrist, Ralph T Mosley, Thomas A Miller
ABSTRACT
Benzo[b]thienyl hydroxamic acids, a novel class of histone deacetylase (HDAC) inhibitors, were identified via a targeted screen of small molecule hydroxamic acids. Various substitutions were explored in the C5- and C6-positions of the benzo[b]thiophene core to characterize SAR and develop optimal inhibitors. It was determined that substitution at the C6-position of the benzo[b]thiophene core with a three-atom spacer yielded optimal HDAC1 inhibition and anti-proliferative activity in murine erythroleukemia (SC-9) cells.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acetohydroxamic acid, 98%
Sigma-Aldrich
Benzhydroxamic acid, 99%