Merck
  • Hypoxia Attenuates Trastuzumab Uptake and Trastuzumab-Emtansine (T-DM1) Cytotoxicity through Redistribution of Phosphorylated Caveolin-1.
Hypoxia Attenuates Trastuzumab Uptake and Trastuzumab-Emtansine (T-DM1) Cytotoxicity through Redistribution of Phosphorylated Caveolin-1.
Molecular cancer research : MCR (2020-01-05)
Vineesh Indira Chandran, Ann-Sofie Månsson, Magdalena Barbachowska, Myriam Cerezo-Magaña, Björn Nodin, Bharat Joshi, Neelima Koppada, Ola M Saad, Oleg Gluz, Karolin Isaksson, Signe Borgquist, Karin Jirström, Ivan Robert Nabi, Helena Jernström, Mattias Belting
ABSTRACT
The antibody-drug conjugate trastuzumab-emtansine (T-DM1) offers an additional treatment option for patients with HER2-amplified tumors. However, primary and acquired resistance is a limiting factor in a significant subset of patients. Hypoxia, a hallmark of cancer, regulates the trafficking of several receptor proteins with potential implications for tumor targeting. Here, we have investigated how hypoxic conditions may regulate T-DM1 treatment efficacy in breast cancer. The therapeutic effect of T-DM1 and its metabolites was evaluated in conjunction with biochemical, flow cytometry, and high-resolution imaging studies to elucidate the functional and mechanistic aspects of hypoxic regulation. HER2 and caveolin-1 expression was investigated in a well-annotated breast cancer cohort. We find that hypoxia fosters relative resistance to T-DM1 in HER2+ cells (SKBR3 and BT474). This effect was not a result of deregulated HER2 expression or resistance to emtansine and its metabolites. Instead, we show that hypoxia-induced translocation of caveolin-1 from cytoplasmic vesicles to the plasma membrane contributes to deficient trastuzumab internalization and T-DM1 chemosensitivity. Caveolin-1 depletion mimicked the hypoxic situation, indicating that vesicular caveolin-1 is indispensable for trastuzumab uptake and T-DM1 cytotoxicity. In vitro studies suggested that HER2 and caveolin-1 are not coregulated, which was supported by IHC analysis in patient tumors. We find that phosphorylation-deficient caveolin-1 inhibits trastuzumab internalization and T-DM1 cytotoxicity, suggesting a specific role for caveolin-1 phosphorylation in HER2 trafficking. IMPLICATIONS: Together, our data for the first time identify hypoxic regulation of caveolin-1 as a resistance mechanism to T-DM1 with potential implications for individualized treatment of breast cancer.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-IgG di topo (molecola intera) - perossidasi, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-c-ErbB2/c-Neu (Ab-5) Mouse mAb (TA-1), liquid, clone TA-1, Calbiochem®