Merck
Peripheral administration of a humanized anti-PrP antibody blocks Alzheimer's disease Aβ synaptotoxicity.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-05-03)
Igor Klyubin, Andrew J Nicoll, Azadeh Khalili-Shirazi, Michael Farmer, Stephanie Canning, Alexandra Mably, Jacqueline Linehan, Alexander Brown, Madeleine Wakeling, Sebastian Brandner, Dominic M Walsh, Michael J Rowan, John Collinge
ABSTRACT
Alzheimer's disease (AD) is associated with pathological assembly states of amyloid-β protein (Aβ). Aβ-related synaptotoxicity can be blocked by anti-prion protein (PrP) antibodies, potentially allowing therapeutic targeting of this aspect of AD neuropathogenesis. Here, we show that intravascular administration of a high-affinity humanized anti-PrP antibody to rats can prevent the plasticity-disrupting effects induced by exposure to soluble AD brain extract. These results provide an in vivo proof of principle for such a therapeutic strategy.