Merck
ELF4/MEF activates MDM2 expression and blocks oncogene-induced p16 activation to promote transformation.
Molecular and cellular biology (2009-04-22)
Goro Sashida, Yan Liu, Shannon Elf, Yasuhiko Miyata, Kazuma Ohyashiki, Miki Izumi, Silvia Menendez, Stephen D Nimer
ABSTRACT
Several ETS transcription factors, including ELF4/MEF, can function as oncogenes in murine cancer models and are overexpressed in human cancer. We found that Elf4/Mef activates Mdm2 expression; thus, lack of or knockdown of Elf4/Mef reduces Mdm2 levels in mouse embryonic fibroblasts (mef's), leading to enhanced p53 protein accumulation and p53-dependent senescence. Even though p53 is absent in Elf4(-/-) p53(-/-) mef's, neither oncogenic H-Ras(V12) nor c-myc can induce transformation of these cells. This appears to relate to the INK4a/ARF locus; both p19(ARF) and p16 are increased in Elf4(-/-) p53(-/-) mef's, and expression of Bmi-1 or knockdown of p16 in this context restores H-Ras(V12)-induced transformation. Thus, ELF4/MEF promotes tumorigenesis by inhibiting both the p53 and p16/Rb pathways.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-p16 (Ab-1) Mouse mAb (DCS-50.1/H4), liquid, clone DCS-50.1/H4, Calbiochem®