Merck
ERdj8 governs the size of autophagosomes during the formation process.
The Journal of cell biology (2020-06-04)
Yo-Hei Yamamoto, Ayano Kasai, Hiroko Omori, Tomoe Takino, Munechika Sugihara, Tetsuo Umemoto, Maho Hamasaki, Tomohisa Hatta, Tohru Natsume, Richard I Morimoto, Ritsuko Arai, Satoshi Waguri, Miyuki Sato, Ken Sato, Shoshana Bar-Nun, Tamotsu Yoshimori, Takeshi Noda, Kazuhiro Nagata
ABSTRACT
In macroautophagy, membrane structures called autophagosomes engulf substrates and deliver them for lysosomal degradation. Autophagosomes enwrap a variety of targets with diverse sizes, from portions of cytosol to larger organelles. However, the mechanism by which autophagosome size is controlled remains elusive. We characterized a novel ER membrane protein, ERdj8, in mammalian cells. ERdj8 localizes to a meshwork-like ER subdomain along with phosphatidylinositol synthase (PIS) and autophagy-related (Atg) proteins. ERdj8 overexpression extended the size of the autophagosome through its DnaJ and TRX domains. ERdj8 ablation resulted in a defect in engulfing larger targets. C. elegans, in which the ERdj8 orthologue dnj-8 was knocked down, could perform autophagy on smaller mitochondria derived from the paternal lineage but not the somatic mitochondria. Thus, ERdj8 may play a critical role in autophagosome formation by providing the capacity to target substrates of diverse sizes for degradation.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anticorpo anti-Atg13, clone 2H4.2, clone 2H4.2, from mouse