Merck
  • A protocol for Agrobacterium-mediated transformation of Japanese cedar, Sugi (Cryptomeria japonica D. Don) using embryogenic tissue explants.
A protocol for Agrobacterium-mediated transformation of Japanese cedar, Sugi (Cryptomeria japonica D. Don) using embryogenic tissue explants.
Plant biotechnology (Tokyo, Japan) (2020-08-22)
Ken-Ichi Konagaya, Yoshihiko Nanasato, Toru Taniguchi
ABSTRACT
Sugi (Cryptomeria japonica D. Don) is the most important afforestation coniferous tree in Japan. Coniferous trees normally have a long juvenile period and require a long cultivation time for breeding. Through a traditional breeding project that began in the 1950s, first generation plus trees with excellent traits were selected primarily from artificial forests and used as seedlings. Recently, the second generation plus trees obtained by crossing between plus trees have been selected. In light of this situation, the improvement of Sugi by a transgenic approach is effective in terms of shortening the breeding period compared with conventional crossing-dependent approaches. There are three key points to an efficient Agrobacterium-mediated transformation system: (1) establishment of explants with high regeneration ability, (2) optimal co-cultivation conditions for explants and Agrobacterium, and (3) efficient elimination of Agrobacterium. Here we describe a protocol for Agrobacterium-mediated transformation of Sugi that meets the above criteria using embryogenic tissues as explants isolated from immature seeds obtained by crossing.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Gomma di Gellan, Gelrite®