Merck
CHD4 Predicts Aggressiveness in PTC Patients and Promotes Cancer Stemness and EMT in PTC Cells.
International journal of molecular sciences (2021-01-10)
Poyil Pratheeshkumar, Abdul K Siraj, Sasidharan Padmaja Divya, Sandeep Kumar Parvathareddy, Khadija Alobaisi, Saif S Al-Sobhi, Fouad Al-Dayel, Khawla S Al-Kuraya
ABSTRACT
Chromodomain-helicase-DNA-binding protein 4 (CHD4), a core subunit of the nucleosome remodeling and deacetylation (NuRD) complex is highly expressed in several cancers. However, its role in the pathogenesis and progression of papillary thyroid carcinoma (PTC) has not been investigated. We investigated the prognostic significance of CHD4 in a large cohort of Middle Eastern PTC patients and explored the functional role of CHD4 in regulating cancer stemness and EMT in PTC cells. CHD4 overexpression was observed in 45.3% (650/1436) of PTCs, and was associated with aggressive clinico-pathological parameters and worse outcome. Functional analysis using PTC cell lines showed that forced expression of CHD4 promoted cell proliferation, spheroid growth, migration, invasion and progression of epithelial to mesenchymal transition (EMT) in PTC cells whereas its knockdown reversed the effect. Methylation of E-cadherin was associated with loss of expression in CHD4 expressing cells, while CHD4 depletion reactivated E-cadherin expression. Most importantly, knockdown of mesenchymal transcriptional factors, Snail1 or Zeb1, attenuated the spheroid growth in CHD4 expressing PTC cells, showing a potential link between EMT activation and stemness maintenance in PTC. These findings suggest that CHD4 might be a promising therapeutic target in the treatment of patients with an aggressive subtype of PTC.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
MISSION® esiRNA, targeting human CHD4